General Description
DNA pcDNA3_verB
Entire molecule length: 5446 bp
Restriction Map
Enzyme |
# of cuts |
Positions |
AatI |
1 |
2069 |
AatII |
5 |
376 429 512 698 5446 |
Acc65I |
1 |
896 |
AccI |
4 |
34 3251 3258 5446 |
AciI |
69 |
62 309 337 349 363 530 621(c) 654(c) 758 779(c) 925
965(c) 969 1252(c) 1276(c) 1325(c) 1339(c) 1342(c) 1370 1397 1918 1930 1939
1951 1961 1972 2018 2189 2252 2346(c) 2410(c) 2511(c) 2514(c) 2754 2794(c)
2799 2849(c) 2865 2891 2947(c) 3016 3019 3085(c) 3321 3424 3480(c) 3490(c)
3514 3557(c) 3564(c) 3585(c) 3676 3704 3831 3850 3971 4081(c) 4216 4225(c)
4587 4678 4869(c) 4915 5036(c) 5080 5157 5266 5365(c) 5412 |
AcsI |
4 |
128 939 1726 3170 |
AcyI |
9 |
373 426 509 695 2280 2982 3061 5061 5443 |
AflIII |
2 |
229 3631 |
AluI |
26 |
136 817 892 904 1024 1289 1456 1713 2034 2088 2098
2386 2844 3135 3268 3274 3296 3391 3455 3573 3799 3935 4192 4713 4813 4876 |
Alw44I |
2 |
3945 5191 |
AlwI |
19 |
13 16(c) 903(c) 916 2124 2148 2459 2524(c) 2705
3068(c) 3097 4192(c) 4278(c) 4280 4376(c) 4377 4840(c) 5157 5161(c) |
AlwNI |
2 |
81 4047 |
AosI |
2 |
2382 4746 |
ApaI |
1 |
994 |
ApaLI |
2 |
3945 5191 |
ApoI |
4 |
128 939 1726 3170 |
AseI |
6 |
258 863 1740 3402 3461 4696 |
AsnI |
6 |
258 863 1740 3402 3461 4696 |
Asp700 |
2 |
1741 5123 |
Asp718 |
1 |
896 |
AspEI |
1 |
4524 |
AspHI |
8 |
819 906 1026 2393 2583 3949 5110 5195 |
AspI |
1 |
2398 |
AsuII |
1 |
2962 |
AvaI |
2 |
972 2091 |
AvaII |
3 |
2796 4662 4884 |
AviII |
2 |
2382 4746 |
AvrII |
1 |
2070 |
BamHI |
1 |
908 |
BanI |
8 |
716 896 1108 1502 2279 2314 3375 4472 |
BanII |
7 |
819 906 994 1026 1261 1297 2645 |
BbsI |
1 |
1232 |
BbvI |
18 |
188(c) 1373 2227(c) 2353 2395 2411(c) 2504(c) 2916
3144 3442(c) 3523(c) 3541(c) 3960(c) 4050 4053 4259 4562(c) 4953 |
BcgI |
1 |
5063 |
BclI |
2 |
1030 2121 |
BfaI |
15 |
251 830 915 985 1019 1052 1298 1390 2071 3205 3265
3269 4126 4379 4714 |
BglI |
4 |
341 463 534 4644 |
BglII |
1 |
13 |
BlnI |
1 |
2070 |
BmyI |
14 |
819 906 994 1026 1261 1297 2226 2319 2393 2583 2645
3949 5110 5195 |
BpmI |
3 |
3063(c) 3120 4594(c) |
BpuAI |
1 |
1232 |
BsaAI |
3 |
591 1543 2584 |
BsaBI |
2 |
953 2139 |
BsaHI |
9 |
373 426 509 695 2280 2982 3061 5061 5443 |
BsaI |
2 |
877(c) 4585(c) |
BsaJI |
13 |
611 1100 1781 1790 1854 1977 2012 2070 2091 2443
2712 3370 3791 |
BsaWI |
4 |
2311 3837 3984 4815 |
BsiEI |
7 |
925 969 2189 3547 3971 4894 5043 |
BsiHKAI |
8 |
819 906 1026 2393 2583 3949 5110 5195 |
BsiYI |
14 |
300 893 1324 1650 1795 1978 2261 2805 3086 3479
3653 3671 3837 4116 |
BslI |
14 |
300 893 1324 1650 1795 1978 2261 2805 3086 3479
3653 3671 3837 4116 |
BsmAI |
5 |
685 877(c) 2122(c) 4585(c) 5361 |
BsmFI |
9 |
426 577 745 1763(c) 1836(c) 1900(c) 2431 2963 3072 |
BsmI |
1 |
3200(c) |
Bsp120I |
1 |
990 |
Bsp1286I |
14 |
819 906 994 1026 1261 1297 2226 2319 2393 2583 2645
3949 5110 5195 |
BspHI |
2 |
4351 5359 |
BspMI |
3 |
2167(c) 2548 2998 |
BspWI |
37 |
198 341 463 495 534 627 651 974 1249 1273 1334 1336
1378 1405 1435 1792 1815 1887 1938 2017 2023 2271 2355 2378 2517 2523 2640
2676 2723 2990 3375 3419 3503 3570 3684 4256 4644 |
BsrBI |
7 |
971(c) 1399(c) 2893(c) 2947 3323(c) 3564 5365 |
BsrDI |
3 |
2513 4585 4759(c) |
BsrFI |
5 |
1438 2599 2780 3063 4604 |
BsrI |
18 |
75(c) 546(c) 853 928(c) 966 1632 1955(c) 2224 2425
3431(c) 4039 4052 4164(c) 4570(c) 4688(c) 4731(c) 5000 5170(c) |
BssHII |
1 |
2677 |
Bst1107I |
1 |
3252 |
BstBI |
1 |
2962 |
BstNI |
12 |
341 534 1102 1783 1792 1839 1856 2667 3371 3659
3780 3793 |
BstUI |
19 |
121 209 231 311 1315 1339 1359 1735 2346 2647 2679
3085 3478 3480 3678 4259 4589 5082 5414 |
BstXI |
2 |
935 961 |
BstYI |
12 |
13 908 2116 2451 2697 3089 4272 4283 4369 4381 5149
5166 |
CfoI |
30 |
121 201 1317 1330 1339 1361 1387 1395 2274 2282
2346 2383 2649 2679 2681 2909 3085 3415 3480 3508 3541 3811 3878 3978 4152
4261 4654 4747 5084 5416 |
Cfr10I |
5 |
1438 2599 2780 3063 4604 |
Csp45I |
1 |
2962 |
Csp6I |
11 |
44 214 469 494 549 582 633 790 897 2585 5003 |
DdeI |
11 |
40 110 181 1162 1271 2030 2943 3906 4315 4481 5021 |
DpnI |
28 |
7 15 23 910 1032 2118 2123 2142 2453 2531 2612 2621
2699 3075 3091 4199 4274 4285 4293 4371 4383 4488 4829 4847 4893 5151 5168
5204 |
DpnII |
28 |
5 13 21 908 1030 2116 2121 2140 2451 2529 2610 2619
2697 3073 3089 4197 4272 4283 4291 4369 4381 4486 4827 4845 4891 5149 5166
5202 |
DraI |
3 |
4390 4409 5101 |
DraII |
2 |
990 991 |
DraIII |
1 |
1546 |
DrdI |
3 |
1590 2307 3739 |
DsaI |
3 |
611 1977 2712 |
DsaV |
20 |
339 532 1100 1781 1790 1837 1854 2090 2091 2282
2442 2665 3055 3369 3657 3778 3791 4009 4705 5056 |
EaeI |
8 |
922 966 2186 2360 2751 2778 3470 4912 |
EagI |
3 |
922 966 2186 |
Eam1105I |
1 |
4524 |
EarI |
4 |
2624(c) 2834(c) 3515 5319 |
Ecl136II |
3 |
817 904 1024 |
EclXI |
3 |
922 966 2186 |
Eco57I |
4 |
2426 2858 4179 5191(c) |
EcoO109I |
2 |
990 991 |
EcoRI |
1 |
939 |
EcoRII |
12 |
339 532 1100 1781 1790 1837 1854 2665 3369 3657
3778 3791 |
EcoRV |
1 |
951 |
Fnu4HI |
40 |
62 202 925 966 969 1326 1340 1362 2018 2189 2241
2252 2342 2347 2384 2425 2512 2515 2518 2754 2850 2891 2905 3019 3133 3456
3537 3555 3558 3676 3831 3974 4039 4042 4248 4576 4915 4942 5037 5266 |
FnuDII |
19 |
121 209 231 311 1315 1339 1359 1735 2346 2647 2679
3085 3478 3480 3678 4259 4589 5082 5414 |
FokI |
10 |
1261 1456(c) 1921(c) 2147 2604 2629 3083 4490(c)
4671(c) 4958(c) |
FspI |
2 |
2382 4746 |
HaeII |
5 |
1388 1396 2283 3509 3879 |
HaeIII |
23 |
220 335 528 924 968 992 1551 1693 2011 2017 2069
2188 2362 2753 2780 3472 3646 3657 3675 4109 4567 4647 4914 |
HgaI |
6 |
777 2990 3069 3742 4320 5050(c) |
HgiAI |
8 |
819 906 1026 2393 2583 3949 5110 5195 |
HhaI |
30 |
121 201 1317 1330 1339 1361 1387 1395 2274 2282
2346 2383 2649 2679 2681 2909 3085 3415 3480 3508 3541 3811 3878 3978 4152
4261 4654 4747 5084 5416 |
HinP1I |
30 |
119 199 1315 1328 1337 1359 1385 1393 2272 2280
2344 2381 2647 2677 2679 2907 3083 3413 3478 3506 3539 3809 3876 3976 4150
4259 4652 4745 5082 5414 |
HincII |
4 |
1 35 235 3259 |
HindII |
4 |
1 35 235 3259 |
HindIII |
1 |
890 |
HinfI |
16 |
36 174 661 870 1591 1613 2765 2899 2951 3009 3046
3466 3531 3606 4002 4519 |
HpaII |
22 |
1439 2092 2185 2262 2284 2312 2443 2533 2600 2781
3056 3064 3349 3838 3985 4011 4201 4605 4639 4706 4816 5058 |
HphI |
9 |
627 1000(c) 1543 2458(c) 4367(c) 4594(c) 5010
5216(c) 5251 |
ItaI |
40 |
62 202 925 966 969 1326 1340 1362 2018 2189 2241
2252 2342 2347 2384 2425 2512 2515 2518 2754 2850 2891 2905 3019 3133 3456
3537 3555 3558 3676 3831 3974 4039 4042 4248 4576 4915 4942 5037 5266 |
KasI |
1 |
2279 |
KpnI |
1 |
900 |
Ksp632I |
4 |
2624(c) 2834(c) 3515 5319 |
MaeI |
15 |
251 830 915 985 1019 1052 1298 1390 2071 3205 3265
3269 4126 4379 4714 |
MaeII |
16 |
373 385 426 509 590 695 1432 1542 1585 1597 2396
2583 4334 4750 5123 5443 |
MaeIII |
18 |
312 399 748 917 1006 1353 1365 2400 2706 3144 3987
4050 4166 4449 4780 4838 4991 5179 |
MamI |
2 |
953 2139 |
MboI |
28 |
5 13 21 908 1030 2116 2121 2140 2451 2529 2610 2619
2697 3073 3089 4197 4272 4283 4291 4369 4381 4486 4827 4845 4891 5149 5166
5202 |
MboII |
13 |
183 1237 1404(c) 2641 2851 2931(c) 3098(c) 3502(c)
4293 4364(c) 5119(c) 5197(c) 5306(c) |
McrI |
7 |
925 969 2189 3547 3971 4894 5043 |
MfeI |
1 |
162 |
MluI |
1 |
229 |
MluNI |
1 |
2362 |
MnlI |
32 |
96(c) 792(c) 981(c) 1047 1089 1134(c) 1209(c)
1266(c) 1516 2001(c) 2007(c) 2031 2037 2044(c) 2047(c) 2059(c) 2131(c)
2195(c) 2331(c) 2688(c) 2881 3087 3272 3480(c) 3530 3739(c) 3813 4063(c)
4463(c) 4544(c) 4691 4897 |
MscI |
1 |
2362 |
MseI |
20 |
70 132 258 863 1333 1604 1702 1719 1730 1740 3402
3461 4337 4389 4394 4408 4461 4696 4735 5100 |
MslI |
6 |
616 1013 2717 4776 4935 5294 |
MspA1I |
6 |
1289 2386 3455 3973 4218 5159 |
MspI |
22 |
1439 2092 2185 2262 2284 2312 2443 2533 2600 2781
3056 3064 3349 3838 3985 4011 4201 4605 4639 4706 4816 5058 |
MunI |
1 |
162 |
MvaI |
12 |
341 534 1102 1783 1792 1839 1856 2667 3371 3659
3780 3793 |
MvnI |
19 |
121 209 231 311 1315 1339 1359 1735 2346 2647 2679
3085 3478 3480 3678 4259 4589 5082 5414 |
MwoI |
37 |
198 341 463 495 534 627 651 974 1249 1273 1334 1336
1378 1405 1435 1792 1815 1887 1938 2017 2023 2271 2355 2378 2517 2523 2640
2676 2723 2990 3375 3419 3503 3570 3684 4256 4644 |
NaeI |
3 |
1440 2782 3065 |
NarI |
1 |
2280 |
NciI |
8 |
2092 2093 2284 2444 3057 4011 4707 5058 |
NcoI |
3 |
611 1977 2712 |
NdeI |
1 |
485 |
NdeII |
28 |
5 13 21 908 1030 2116 2121 2140 2451 2529 2610 2619
2697 3073 3089 4197 4272 4283 4291 4369 4381 4486 4827 4845 4891 5149 5166
5202 |
NgoMI |
3 |
1438 2780 3063 |
NlaIII |
23 |
171 555 615 981 1243 1818 1890 1981 2154 2499 2685
2716 2742 3098 3246 3289 3635 4355 4846 4856 4934 4970 5363 |
NlaIV |
19 |
718 898 910 992 1110 1483 1504 1787 1860 2281 2316
3377 3663 3702 4474 4568 4609 4820 5410 |
NotI |
1 |
966 |
NruI |
1 |
209 |
NsiI |
3 |
983 1820 1892 |
NspI |
6 |
981 1243 1818 1890 2685 3635 |
NspV |
1 |
2962 |
PaeR7I |
1 |
972 |
PleI |
9 |
30(c) 655(c) 864(c) 1599 1607(c) 2945(c) 3525(c)
4010 4513(c) |
Ppu10I |
3 |
979 1816 1888 |
Psp1406I |
2 |
4750 5123 |
PstI |
2 |
948 2333 |
PvuI |
1 |
4894 |
PvuII |
3 |
1289 2386 3455 |
RcaI |
2 |
4351 5359 |
RsaI |
11 |
45 215 470 495 550 583 634 791 898 2586 5004 |
RsrII |
1 |
2796 |
SacI |
3 |
819 906 1026 |
SalI |
3 |
33 3257 5445 |
SapI |
3 |
2624(c) 2834(c) 3515 |
Sau3AI |
28 |
5 13 21 908 1030 2116 2121 2140 2451 2529 2610 2619
2697 3073 3089 4197 4272 4283 4291 4369 4381 4486 4827 4845 4891 5149 5166
5202 |
Sau96I |
11 |
218 334 527 990 991 1549 2796 4566 4645 4662 4884 |
ScaI |
1 |
5004 |
ScrFI |
20 |
341 534 1102 1783 1792 1839 1856 2092 2093 2284
2444 2667 3057 3371 3659 3780 3793 4011 4707 5058 |
SexAI |
1 |
1837 |
SfaNI |
19 |
48(c) 608(c) 990 1159 1239(c) 1478 1827 1899
2238(c) 2493(c) 2579 2643 2709(c) 2918 3172 3728 4780 4971(c) 5220 |
SfcI |
8 |
876 944 1000 1320 2329 3896 4087 4765 |
SfuI |
1 |
2962 |
SmaI |
1 |
2093 |
SnaBI |
1 |
591 |
SnoI |
2 |
3945 5191 |
SpeI |
2 |
250 914 |
SphI |
5 |
981 1243 1818 1890 2685 |
SspI |
1 |
5328 |
StuI |
1 |
2069 |
StyI |
4 |
611 1977 2070 2712 |
TaqI |
16 |
34 857 973 1039 1508 2393 2549 2573 2609 2771 2962
3007 3258 3731 5175 5446 |
TfiI |
7 |
174 2765 2899 3009 3046 3466 3606 |
ThaI |
19 |
121 209 231 311 1315 1339 1359 1735 2346 2647 2679
3085 3478 3480 3678 4259 4589 5082 5414 |
Tru9I |
20 |
70 132 258 863 1333 1604 1702 1719 1730 1740 3402
3461 4337 4389 4394 4408 4461 4696 4735 5100 |
Tsp509I |
19 |
128 162 269 860 939 1145 1726 1737 1741 1824 1896
1988 3170 3311 3328 3403 4391 4697 4952 |
Tth111I |
1 |
2398 |
XbaI |
1 |
984 |
XhoI |
1 |
972 |
XhoII |
12 |
13 908 2116 2451 2697 3089 4272 4283 4369 4381 5149
5166 |
XmaI |
1 |
2091 |
XmaIII |
3 |
922 966 2186 |
XmnI |
2 |
1741 5123 |
No
cuts: AccIII, AflII, AgeI, AscI, BbrPI, BfrI, Bpu1102I, BseAI, BsgI, BsiWI,
BspDI, BspEI, BsrGI, BstEII, Bsu36I, CelII, ClaI, Eco47III, EcoNI, Esp3I, EspI,
HpaI, KspI, MroI, NheI, PacI, PflMI, PinAI, PmaCI, PmeI, PmlI, PpuMI, SacII,
SfiI, SgrAI, SspBI, SwaI, Van91I, XcmI