General Description
DNA pLysE
Entire molecule length: 4886 bp
Restriction Map
Enzyme |
# of cuts |
Positions |
AccI |
2 |
596 2789 |
AccIII |
2 |
6 4080 |
AciI |
57 |
26(c) 569 588(c) 708 751(c) 758(c) 780(c) 833
835(c) 861 952(c) 1005 1017 1110 1328 1340(c) 1534(c) 1676(c) 1784 1793 1796
2404(c) 2562(c) 2715(c) 2718(c) 2721(c) 2840(c) 2860 2939 2979(c) 3028
3075(c) 3160(c) 3175 3243(c) 3276 3301 3346 3370 3382(c) 3424(c) 3478(c) 3531
3547 3553(c) 3634(c) 3772(c) 3803 3867(c) 3900 3905 3934(c) 3954 4309(c) 4379
4454 4548(c) |
AcsI |
3 |
2 4350 4362 |
AcyI |
6 |
2154 2297 2551 2572 2686 3343 |
AflII |
1 |
2324 |
AgeI |
4 |
670 993 3824 4284 |
AluI |
13 |
106 225 234 518 535 1074 1307 1511 1526 2824 3227
3918 4863 |
AlwI |
6 |
1865(c) 1878 2507(c) 2520 3242 4091 |
AlwNI |
3 |
538 1186 4166 |
AosI |
4 |
1757 2441 3495 3593 |
ApoI |
3 |
2 4350 4362 |
AseI |
2 |
1407 2375 |
AsnI |
2 |
1407 2375 |
Asp700 |
1 |
640 |
AspEI |
1 |
3851 |
AspHI |
5 |
495 1775 2728 3315 3606 |
AspI |
1 |
4344 |
AsuII |
2 |
1320 4360 |
AvaI |
1 |
3562 |
AvaII |
6 |
1877 2936 3024 3273 3576 3618 |
AviII |
4 |
1757 2441 3495 3593 |
BamHI |
2 |
1870 2512 |
BanI |
11 |
1571 1614 2296 2550 2571 2685 2903 3342 3426 3997
4422 |
BanII |
2 |
2612 2626 |
BbsI |
4 |
2267(c) 2867(c) 3730(c) 3961(c) |
BbvI |
16 |
437(c) 652(c) 1099(c) 1189 1192 1708(c) 2510
2739(c) 2922 3555 3579 3683(c) 3686(c) 3776(c) 4016 4088(c) |
BcgI |
3 |
2210(c) 2834(c) 3928(c) |
BclI |
1 |
4184 |
BfaI |
4 |
585 1426 1725 3626 |
BfrI |
1 |
2324 |
BglI |
2 |
3072 3306 |
BmyI |
8 |
495 1775 2612 2626 2728 3315 3606 4425 |
BpmI |
4 |
479(c) 2969 3523(c) 4772(c) |
BpuAI |
4 |
2267(c) 2867(c) 3730(c) 3961(c) |
BsaAI |
2 |
313 3786 |
BsaBI |
1 |
4088 |
BsaHI |
6 |
2154 2297 2551 2572 2686 3343 |
BsaI |
1 |
2389(c) |
BsaJI |
20 |
349 350 833 947 1286 1610 1624 2365 2471 2665 2671
3304 3506 3584 4020 4026 4208 4587 4656 4657 |
BsaWI |
11 |
6 670 993 1123 2830 3824 3911 3924 4080 4269 4284 |
BseAI |
2 |
6 4080 |
BsgI |
2 |
651(c) 4051(c) |
BsiEI |
6 |
741 1110 1784 2793 3079 4343 |
BsiHKAI |
5 |
495 1775 2728 3315 3606 |
BsiYI |
22 |
344 1676 1767 1867 2290 2491 2670 2671 2717 2747
2765 2833 3458 3507 3585 3911 3927 4027 4028 4211 4342 4663 |
BslI |
22 |
344 1676 1767 1867 2290 2491 2670 2671 2717 2747
2765 2833 3458 3507 3585 3911 3927 4027 4028 4211 4342 4663 |
BsmAI |
5 |
331 1030 2070(c) 2389(c) 4664(c) |
BsmFI |
8 |
386 506(c) 1863(c) 2689 3010(c) 3235 3763(c)
4002(c) |
BsmI |
3 |
15(c) 3496 4494(c) |
Bsp1286I |
8 |
495 1775 2612 2626 2728 3315 3606 4425 |
BspDI |
1 |
1519 |
BspEI |
2 |
6 4080 |
BspHI |
2 |
2287 2626 |
BspMI |
1 |
3191(c) |
BspWI |
39 |
231 435 499 764 1013 1174 1661 1727 1734 1792 1814
1848 2032 2041 2303 2548 2559 2580 2771 2912 2938 3063 3072 3081 3179 3183
3188 3197 3297 3306 3555 3564 3589 3631 3696 3719 3798 4006 4554 |
BsrBI |
2 |
758 3867 |
BsrDI |
1 |
4875(c) |
BsrFI |
12 |
670 993 1226 1655 2538 2547 2906 3066 3420 3824
4000 4284 |
BsrGI |
1 |
1927 |
BsrI |
17 |
201(c) 496(c) 604 1178 1191 1279(c) 1705(c) 1800(c)
2020(c) 2355 2752 2817(c) 3026 3275 3652 4234 4647 |
Bst1107I |
1 |
597 |
BstBI |
2 |
1320 4360 |
BstNI |
12 |
351 379 791 802 937 949 1626 3197 3580 4410 4658
4714 |
BstUI |
19 |
835 1019 1372 1842 2280 2840 2955 3084 3111 3116
3177 3243 3372 3382 3527 3553 3675 3772 3934 |
BstYI |
3 |
1870 2512 4083 |
Bsu36I |
1 |
4235 |
CfoI |
30 |
431 584 967 1117 1226 1268 1374 1598 1730 1758 2270
2282 2299 2442 2553 2574 2634 2688 2840 2915 2955 3086 3345 3496 3558 3594
3934 4009 4063 4146 |
Cfr10I |
12 |
670 993 1226 1655 2538 2547 2906 3066 3420 3824
4000 4284 |
ClaI |
1 |
1519 |
Csp45I |
2 |
1320 4360 |
Csp6I |
6 |
126 1659 1928 2082 2260 4474 |
DdeI |
11 |
230 1045 1308 2127 2256 3718 3848 3885 4221 4235
4668 |
DpnI |
16 |
277 320 404 1387 1398 1845 1872 2514 2605 2964 3236
3267 3282 3599 4085 4186 |
DpnII |
16 |
275 318 402 1385 1396 1843 1870 2512 2603 2962 3234
3265 3280 3597 4083 4184 |
DraI |
3 |
86 1936 4633 |
DraII |
5 |
1877 2661 3576 3618 4031 |
DrdI |
1 |
4344 |
DsaI |
7 |
833 2365 2665 3584 4026 4208 4587 |
DsaV |
23 |
266 349 358 377 789 800 935 947 1051 1148 1624 1664
1873 2670 3195 3394 3578 3620 4020 4342 4408 4656 4712 |
EaeI |
8 |
1014 1790 2536 2668 3076 3581 4023 4623 |
EagI |
1 |
3076 |
Eam1105I |
1 |
3851 |
EarI |
1 |
1453 |
EclXI |
1 |
3076 |
Eco47III |
5 |
583 1729 2633 2914 4145 |
Eco57I |
2 |
1485 3814 |
EcoNI |
1 |
2763 |
EcoO109I |
5 |
1877 2661 3576 3618 4031 |
EcoRI |
1 |
2 |
EcoRII |
12 |
349 377 789 800 935 947 1624 3195 3578 4408 4656
4712 |
EcoRV |
1 |
1682 |
Esp3I |
3 |
331 2070(c) 4664(c) |
Fnu4HI |
38 |
451 666 752 833 836 953 1017 1113 1178 1181 1722
1793 1796 2499 2716 2719 2753 2860 2911 3076 3161 3244 3301 3346 3425 3544
3547 3554 3568 3697 3700 3790 3868 3954 4005 4102 4310 4549 |
FnuDII |
19 |
835 1019 1372 1842 2280 2840 2955 3084 3111 3116
3177 3243 3372 3382 3527 3553 3675 3772 3934 |
FokI |
7 |
22 1593(c) 1641 3137 3182 4110 4362(c) |
FspI |
4 |
1757 2441 3495 3593 |
HaeII |
13 |
585 1731 2271 2300 2554 2575 2635 2689 2916 3346
4010 4064 4147 |
HaeIII |
25 |
48 93 348 832 1016 1669 1792 2509 2538 2662 2670
2734 2968 3057 3078 3129 3186 3399 3583 3870 4025 4033 4233 4625 4712 |
HgaI |
12 |
427(c) 665(c) 899 1964 2143(c) 2204 2536 2775(c)
3090 3122 3366(c) 3516(c) |
HgiAI |
5 |
495 1775 2728 3315 3606 |
HhaI |
30 |
431 584 967 1117 1226 1268 1374 1598 1730 1758 2270
2282 2299 2442 2553 2574 2634 2688 2840 2915 2955 3086 3345 3496 3558 3594
3934 4009 4063 4146 |
HinP1I |
30 |
429 582 965 1115 1224 1266 1372 1596 1728 1756 2268
2280 2297 2440 2551 2572 2632 2686 2838 2913 2953 3084 3343 3494 3556 3592
3932 4007 4061 4144 |
HincII |
2 |
2790 3856 |
HindII |
2 |
2790 3856 |
HindIII |
1 |
1524 |
HinfI |
11 |
725 1141 1946 2014 2769 2989 3143 3441 3662 3852
4539 |
HpaII |
36 |
7 49 268 360 572 671 994 1052 1124 1150 1227 1656
1665 1874 2510 2524 2539 2548 2671 2831 2907 3067 3157 3395 3421 3622 3825
3912 3925 4001 4022 4081 4270 4285 4343 4765 |
HphI |
16 |
304 456 1271 1613(c) 2537(c) 2582(c) 3436(c)
3657(c) 3839 4603 4645(c) 4657 4713(c) 4849(c) 4851 4857(c) |
ItaI |
38 |
451 666 752 833 836 953 1017 1113 1178 1181 1722
1793 1796 2499 2716 2719 2753 2860 2911 3076 3161 3244 3301 3346 3425 3544
3547 3554 3568 3697 3700 3790 3868 3954 4005 4102 4310 4549 |
KasI |
5 |
2296 2550 2571 2685 3342 |
Ksp632I |
1 |
1453 |
KspI |
1 |
836 |
MaeI |
4 |
585 1426 1725 3626 |
MaeII |
10 |
312 324 1942 3038 3094 3683 3707 3785 4628 4803 |
MaeIII |
16 |
410 553 1126 1259 1281 1619 1707 2132 2242 2353
3017 3284 3888 4257 4725 4830 |
MamI |
1 |
4088 |
MboI |
16 |
275 318 402 1385 1396 1843 1870 2512 2603 2962 3234
3265 3280 3597 4083 4184 |
MboII |
13 |
397(c) 806 817 1406 1440(c) 2267(c) 2353 2613
2867(c) 3138(c) 3730(c) 3961(c) 4618 |
McrI |
6 |
741 1110 1784 2793 3079 4343 |
MluNI |
2 |
3583 4625 |
MnlI |
34 |
161 316 481(c) 534 617(c) 724 821(c) 969 1195(c)
1281 1296 1620 1680 1899(c) 2083(c) 2284(c) 2420(c) 2466(c) 2469(c) 2488(c)
2526 2745 2927(c) 3012 3111(c) 3314 3375 3413 3440 3609(c) 3881 4164(c) 4215
4244 |
MroI |
2 |
6 4080 |
MscI |
2 |
3583 4625 |
MseI |
16 |
85 813 1231 1407 1529 1551 1893 1935 2325 2375 4136
4317 4417 4438 4490 4632 |
MslI |
4 |
290 3168 3599 4073 |
MspA1I |
9 |
106 518 835 1007 1112 2404 3278 3907 4548 |
MspI |
36 |
7 49 268 360 572 671 994 1052 1124 1150 1227 1656
1665 1874 2510 2524 2539 2548 2671 2831 2907 3067 3157 3395 3421 3622 3825
3912 3925 4001 4022 4081 4270 4285 4343 4765 |
MvaI |
12 |
351 379 791 802 937 949 1626 3197 3580 4410 4658
4714 |
MvnI |
19 |
835 1019 1372 1842 2280 2840 2955 3084 3111 3116
3177 3243 3372 3382 3527 3553 3675 3772 3934 |
MwoI |
39 |
231 435 499 764 1013 1174 1661 1727 1734 1792 1814
1848 2032 2041 2303 2548 2559 2580 2771 2912 2938 3063 3072 3081 3179 3183
3188 3197 3297 3306 3555 3564 3589 3631 3696 3719 3798 4006 4554 |
NaeI |
5 |
2540 2908 3068 3422 4002 |
NarI |
5 |
2297 2551 2572 2686 3343 |
NciI |
11 |
268 360 1053 1150 1666 1875 2672 3396 3622 4022
4344 |
NcoI |
1 |
4587 |
NdeII |
16 |
275 318 402 1385 1396 1843 1870 2512 2603 2962 3234
3265 3280 3597 4083 4184 |
NgoMI |
5 |
2538 2906 3066 3420 4000 |
NheI |
2 |
584 1724 |
NlaIII |
24 |
648 1162 1224 1503 1850 2291 2630 2703 2849 2888
3074 3191 3319 3334 3391 3598 3732 3865 3943 3982 4509 4530 4591 4821 |
NlaIV |
28 |
304 859 956 1573 1616 1826 1872 1878 1879 2298 2469
2514 2552 2573 2663 2687 2905 3026 3344 3393 3428 3463 3577 3620 3819 3999
4032 4424 |
NruI |
1 |
3111 |
NspI |
2 |
1162 2703 |
NspV |
2 |
1320 4360 |
PflMI |
4 |
344 3458 3507 4663 |
PinAI |
4 |
670 993 3824 4284 |
PleI |
4 |
719(c) 1149 2777 3860 |
PmeI |
1 |
1936 |
PpuMI |
3 |
1877 3576 3618 |
Psp1406I |
2 |
3038 4803 |
PvuII |
2 |
106 518 |
RcaI |
2 |
2287 2626 |
RsaI |
6 |
127 1660 1929 2083 2261 4475 |
SacII |
1 |
836 |
SalI |
1 |
2788 |
Sau3AI |
16 |
275 318 402 1385 1396 1843 1870 2512 2603 2962 3234
3265 3280 3597 4083 4184 |
Sau96I |
13 |
347 830 1667 1877 2507 2661 2936 3024 3273 3397
3576 3618 4031 |
ScaI |
1 |
4475 |
ScrFI |
23 |
268 351 360 379 791 802 937 949 1053 1150 1626 1666
1875 2672 3197 3396 3580 3622 4022 4344 4410 4658 4714 |
SfaNI |
16 |
159(c) 437(c) 787(c) 885 1619(c) 1708 1732(c) 2539
2551 2785(c) 3160(c) 3172 3567 4088(c) 4098 4560 |
SfcI |
2 |
1633 2112 |
SfuI |
2 |
1320 4360 |
SgrAI |
2 |
670 2547 |
SphI |
1 |
2703 |
SspBI |
1 |
1927 |
SspI |
2 |
1421 4580 |
StyI |
2 |
3506 4587 |
TaqI |
14 |
247 745 1320 1519 1834 2046 2175 2193 2319 2789
3264 3405 4348 4360 |
TfiI |
7 |
1946 2014 2989 3143 3441 3662 4539 |
ThaI |
19 |
835 1019 1372 1842 2280 2840 2955 3084 3111 3116
3177 3243 3372 3382 3527 3553 3675 3772 3934 |
Tru9I |
16 |
85 813 1231 1407 1529 1551 1893 1935 2325 2375 4136
4317 4417 4438 4490 4632 |
Tsp509I |
16 |
2 1191 1439 1493 1553 1746 2372 2376 2456 3456 3470
4196 4350 4362 4444 4483 |
Tth111I |
1 |
4344 |
Van91I |
4 |
344 3458 3507 4663 |
XbaI |
1 |
1425 |
XhoII |
3 |
1870 2512 4083 |
XmaIII |
1 |
3076 |
XmnI |
1 |
640 |
No
cuts: AatI, AatII, Acc65I, AflIII, Alw44I, ApaI, ApaLI, AscI, Asp718, AvrII,
BbrPI, BglII, BlnI, Bpu1102I, BsiWI, Bsp120I, BssHII, BstEII, BstXI, CelII,
DraIII, Ecl136II, EspI, HpaI, KpnI, MfeI, MluI, MunI, NdeI, NotI, NsiI, PacI,
PaeR7I, PmaCI, PmlI, Ppu10I, PstI, PvuI, RsrII, SacI, SapI, SexAI, SfiI, SmaI,
SnaBI, SnoI, SpeI, StuI, SwaI, XcmI, XhoI, XmaI